siRNA Details
| Gene Symbol: Fam20b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145413.4 | NP_663388.1 | 2 | 527 | ||
| GenBank mRNA | AK034081.1 | - | 3 | 634 | ||
| AK049705.1 | - | 1 | 329 | |||
| AK132149.1 | - | 2 | 528 | |||
| BC019381.1 | - | 2 | 546 | 4206622 | ||
| BC023737.1 | - | 3 | 743 | 5352660 | ||
| BC024412.1 | - | 1 | 477 | 4971483 | ||
| BC031473.1 | - | 2 | 639 | 3990487 | ||

Fam20b
Genomic Map