siRNA Details
| Gene Symbol: Bud13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146000.2 | NP_666112.1 | 10 | 1852 | ||
| GenBank mRNA | AK038126.1 | - | 10 | 1857 | ||
| AK077744.1 | - | 10 | 1857 | |||
| BC025490.1 | - | 10 | 1853 | 5252666 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bud13| Gene Symbol: Bud13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146000.2 | NP_666112.1 | 10 | 1852 | ||
| GenBank mRNA | AK038126.1 | - | 10 | 1857 | ||
| AK077744.1 | - | 10 | 1857 | |||
| BC025490.1 | - | 10 | 1853 | 5252666 | ||