siRNA Details
| Gene Symbol: Slc43a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199283.1 | NP_001186212.1 | 9 | 1382 | ||
| NM_001199284.1 | NP_001186213.1 | 9 | 1164 | |||
| NM_173388.2 | NP_775564.1 | 9 | 1127 | |||
| GenBank mRNA | AB120363.1 | - | 9 | 1160 | ||
| AK143773.1 | - | 9 | 1225 | |||
| AK159144.1 | - | 9 | 1350 | |||
| AK171104.1 | - | 9 | 1384 | |||
| BC042513.1 | - | 9 | 1123 | 3964020 | ||

Slc43a2
Genomic Map