siRNA Details
| Gene Symbol: Ncaph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144818.3 | NP_659067.2 | 2 | 310 | ||
| GenBank mRNA | AK028919.1 | - | 2 | 295 | ||
| AK042706.1 | - | 2 | 227 | |||
| AK080433.1 | - | 2 | 299 | |||
| BC021499.1 | - | 2 | 284 | 5346298 | ||
| BC025060.1 | - | 2 | 235 | 3673890 | ||
| BY772780.1 | - | 2 | 312 | |||

Ncaph
Genomic Map