siRNA Details
| Gene Symbol: Ncaph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144818.3 | NP_659067.2 | 13 | 1684 | ||
| GenBank mRNA | AK028919.1 | - | 13 | 1669 | ||
| AK042706.1 | - | 13 | 1601 | |||
| AK045239.1 | - | 1 | 1033 | |||
| AK080433.1 | - | 13 | 1673 | |||
| AK129047.1 | - | 10 | 1237 | |||
| AK193653.1 | - | 4 | 319 | |||
| BC021499.1 | - | 13 | 1658 | 5346298 | ||
| BC025060.1 | - | 13 | 1609 | 3673890 | ||

Ncaph
Genomic Map