siRNA Details
| Gene Symbol: Cdh12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008420.2 | NP_001008420.1 | 12 | 2166 | ||
| GenBank mRNA | BC086655.1 | - | 12 | 2166 | 30542783 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh12| Gene Symbol: Cdh12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008420.2 | NP_001008420.1 | 12 | 2166 | ||
| GenBank mRNA | BC086655.1 | - | 12 | 2166 | 30542783 | |