siRNA Details
| Gene Symbol: Nav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173437.2 | NP_775613.2 | 3, 4 | 1279 | ||
| GenBank mRNA | AF307453.1 | - | 3, 4 | 1639 | ||
| AK122458.1 | - | 5, 6 | 1517 | |||
| AK140435.1 | - | 3, 4 | 1254 | |||
| BC059840.1 | - | 3, 4 | 1425 | 6836774 | ||

Nav1
Genomic Map