siRNA Details
| Gene Symbol: Nav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173437.2 | NP_775613.2 | 6 | 1849 | ||
| GenBank mRNA | AF307453.1 | - | 6 | 2209 | ||
| AK053255.1 | - | 4 | 828 | |||
| AK082667.1 | - | 4 | 762 | |||
| AK122458.1 | - | 8 | 2087 | |||
| AK140435.1 | - | 6 | 1824 | |||
| BC059840.1 | - | 6 | 1995 | 6836774 | ||

Nav1
Genomic Map