siRNA Details
| Gene Symbol: Adgrg6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002268.3 | NP_001002268.1 | 3 | 294 | ||
| GenBank mRNA | AB183545.1 | - | 3 | 295 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adgrg6| Gene Symbol: Adgrg6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002268.3 | NP_001002268.1 | 3 | 294 | ||
| GenBank mRNA | AB183545.1 | - | 3 | 295 | ||
Set Membership: |
|