siRNA Details
| Gene Symbol: Gm4794 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101452.1 | NP_001094922.1 | 2 | 162 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4794| Gene Symbol: Gm4794 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101452.1 | NP_001094922.1 | 2 | 162 | ||