siRNA Details
| Gene Symbol: Eea1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001932.3 | NP_001001932.1 | 25 | 3905 | ||
| GenBank mRNA | BC075637.1 | - | 25 | 3818 | 30545051 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eea1| Gene Symbol: Eea1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001932.3 | NP_001001932.1 | 25 | 3905 | ||
| GenBank mRNA | BC075637.1 | - | 25 | 3818 | 30545051 | |