siRNA Details
| Gene Symbol: Alx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172553.4 | NP_766141.1 | 4 | 844 | ||
| GenBank mRNA | AK047846.1 | - | 4 | 845 | ||
| AK081538.1 | - | 3 | 269 | |||
| AK086731.1 | - | 4 | 657 | |||
| BC052200.1 | - | 4 | 826 | 6513129 | ||

Alx1
Genomic Map