siRNA Details
| Gene Symbol: Zfc3h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033261.2 | NP_001028433.2 | 29 | 5520 | ||
| GenBank mRNA | AK133182.1 | - | 1 | 111 | ||
| AK161448.1 | - | 1 | 89 | |||
| AK172969.1 | - | 27 | 4220 | |||
| BC033596.1 | - | 7 | 850 | |||
| BC046488.1 | - | 15 | 2365 | |||
| BC061510.1 | - | 17 | 2774 | |||
| BC139124.1 | - | 29 | 5377 | 8862146 | ||

Zfc3h1
Genomic Map