siRNA Details
| Gene Symbol: Zfc3h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033261.2 | NP_001028433.2 | 24 | 4835 | ||
| GenBank mRNA | AK172969.1 | - | 22 | 3535 | ||
| BC033596.1 | - | 2 | 165 | |||
| BC046488.1 | - | 10 | 1680 | |||
| BC061510.1 | - | 12 | 2089 | |||
| BC139124.1 | - | 24 | 4692 | 8862146 | ||

Zfc3h1
Genomic Map