siRNA Details
| Gene Symbol: Tmem5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153059.1 | NP_694699.1 | 3 | 395 | ||
| GenBank mRNA | AK043098.1 | - | 429 | |||
| AK075928.1 | - | 3 | 386 | |||
| AK078061.1 | - | 3 | 463 | |||
| BC020100.1 | - | 3 | 395 | 3981114 | ||
| BC083125.1 | - | 4 | 563 | |||

Tmem5
Genomic Map