siRNA Details
| Gene Symbol: Ccdc157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164620.1 | NP_001158092.1 | 5, 6 | 1354 | ||
| NM_177616.3 | NP_808284.2 | 6, 7 | 1490 | |||
| GenBank mRNA | AK076934.1 | - | 6, 7 | 1418 | ||
| AK133165.1 | - | 5, 6 | 1355 | |||
| AK143216.1 | - | 4, 5 | 1009 | |||
| AK220409.1 | - | 6, 7 | 1471 | |||
| BC120737.1 | - | 3, 4 | 852 | 40129660 | ||
| BC120739.1 | - | 3, 4 | 852 | 40129662 | ||

Ccdc157
Genomic Map