siRNA Details
| Gene Symbol: Ccdc157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164620.1 | NP_001158092.1 | 10 | 2007 | ||
| NM_177616.3 | NP_808284.2 | 11 | 2143 | |||
| GenBank mRNA | AK076934.1 | - | 11 | 2071 | ||
| AK133165.1 | - | 10 | 2008 | |||
| AK143216.1 | - | 9 | 1663 | |||
| AK220409.1 | - | 11 | 2124 | |||
| BC046392.1 | - | 4 | 886 | |||
| BC120737.1 | - | 8 | 1505 | 40129660 | ||
| BC120739.1 | - | 8 | 1505 | 40129662 | ||

Ccdc157
Genomic Map