siRNA Details
| Gene Symbol: Ehbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252515.1 | NP_001239444.1 | 13, 14 | 1955 | ||
| NM_153078.4 | NP_694718.4 | 13, 14 | 1880 | |||
| GenBank mRNA | AF424697.1 | - | 12, 13 | 1730 | ||
| AK173055.1 | - | 13, 14 | 1956 | |||

Ehbp1
Genomic Map