siRNA Details
| Gene Symbol: Ehbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252515.1 | NP_001239444.1 | 25 | 4073 | ||
| NM_153078.4 | NP_694718.4 | 25 | 3998 | |||
| GenBank mRNA | AF424697.1 | - | 24 | 3848 | ||
| AK173055.1 | - | 25 | 4074 | |||
| BC037023.1 | - | 12 | 1625 | |||

Ehbp1
Genomic Map