siRNA Details
| Gene Symbol: Ccdc85a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166661.2 | NP_001160133.1 | 1 | 866 | ||
| NM_181577.5 | NP_853555.2 | 1 | 866 | |||
| GenBank mRNA | AK045568.1 | - | 1 | 839 | ||
| AK087049.1 | - | 1 | 809 | |||
| AK087070.1 | - | 1 | 809 | |||
| AK173298.1 | - | 1 | 883 | |||
| BC066065.1 | - | 1 | 389 | 6850852 | ||

Ccdc85a
Genomic Map