siRNA Details
Gene Symbol: Rufy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172557.2 | NP_766145.1 | 4 | 648 | ||
GenBank mRNA | AJ250024.1 | - | 3 | 324 | ||
AK046625.1 | - | 4 | 665 | |||
AK046633.1 | - | 4 | 649 | |||
AK051178.1 | - | 4 | 654 |
