siRNA Details
| Gene Symbol: Mrpl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175001.3 | NP_778166.2 | 7 | 598 | ||
| GenBank mRNA | AK077615.1 | - | 7 | 592 | ||
| AK086913.1 | - | 7 | 577 | |||
| AK164417.1 | - | 7 | 577 | |||
| AW986653.1 | - | 1 | 152 | |||
| BC061191.1 | - | 7 | 588 | 6703228 | ||

Mrpl22
Genomic Map