siRNA Details
| Gene Symbol: Tom1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039092.3 | NP_001034181.1 | 11 | 1282 | ||
| NM_001039093.1 | NP_001034182.1 | 11 | 1282 | |||
| NM_153080.2 | NP_694720.2 | 11 | 1282 | |||
| GenBank mRNA | AF467887.1 | - | 11 | 1262 | ||
| AK043095.1 | - | 11 | 1262 | |||
| AK054531.1 | - | 11 | 1250 | |||
| AK166040.1 | - | 11 | 1282 | |||
| AK210353.1 | - | 2 | 172 | |||
| AK213329.1 | - | 2 | 172 | |||
| BC062947.1 | - | 11 | 1246 | 30536491 | ||
| BC127266.1 | - | 11 | 1248 | 30357295 | ||

Tom1l2
Genomic Map