siRNA Details
| Gene Symbol: Dhrs7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172112.1 | NP_001165583.1 | 4 | 514 | ||
| NM_145428.2 | NP_663403.1 | 4 | 485 | |||
| GenBank mRNA | AK052209.1 | - | 4 | 437 | ||
| AK139084.1 | - | 4 | 466 | |||
| AK144057.1 | - | 4 | 420 | |||
| BC003479.1 | - | 4 | 440 | 2655855 | ||
| JX196872.1 | - | 4 | 404 | |||

Dhrs7b
Genomic Map