siRNA Details
| Gene Symbol: Usp43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291049.1 | NP_001277978.1 | 15 | 3244 | ||
| NM_173754.4 | NP_776115.2 | 15 | 3259 | |||
| GenBank mRNA | AK042217.1 | - | 15 | 3245 | ||
| AK047339.1 | - | 3 | 1231 | |||
| AK083271.1 | - | 15 | 3097 | |||

Usp43
Genomic Map