siRNA Details
| Gene Symbol: Usp43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291049.1 | NP_001277978.1 | 6 | 1107 | ||
| NM_173754.4 | NP_776115.2 | 6 | 1107 | |||
| GenBank mRNA | AK042217.1 | - | 6 | 1108 | ||
| AK083271.1 | - | 6 | 945 | |||
| BC021474.1 | - | 5 | 397 | 5343429 | ||

Usp43
Genomic Map