siRNA Details
| Gene Symbol: Tecta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009347.2 | NP_033373.2 | 7 | 1193 | ||
| GenBank mRNA | BC138426.1 | - | 7 | 991 | 8861446 | |
| BC171953.1 | - | 7 | 991 | 9054239 | ||
| X99805.1 | - | 7 | 1193 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tecta| Gene Symbol: Tecta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009347.2 | NP_033373.2 | 7 | 1193 | ||
| GenBank mRNA | BC138426.1 | - | 7 | 991 | 8861446 | |
| BC171953.1 | - | 7 | 991 | 9054239 | ||
| X99805.1 | - | 7 | 1193 | |||