siRNA Details
| Gene Symbol: Kdm6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017426.1 | NP_001017426.1 | 21 | 5122 | ||
| GenBank mRNA | AK129121.1 | - | 11 | 1914 | ||
| AK136019.1 | - | 11 | 1303 | |||
| BC029201.1 | - | 2 | 216 | |||
| BC038313.1 | - | 6 | 760 | |||
| BC075632.1 | - | 22 | 5298 | 30362899 | ||

Kdm6b
Genomic Map