siRNA Details
| Gene Symbol: Wscd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177618.4 | NP_808286.2 | 5 | 1200 | ||
| GenBank mRNA | AK220346.1 | - | 4 | 906 | ||
| BC030477.1 | - | 5 | 1337 | |||
| BC048831.1 | - | 4 | 1019 | 4504495 | ||
| BC063070.1 | - | 5 | 1200 | 5687975 | ||

Wscd1
Genomic Map