siRNA Details
| Gene Symbol: Wscd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177618.4 | NP_808286.2 | 7, 8 | 1569 | ||
| GenBank mRNA | AK220346.1 | - | 6, 7 | 1275 | ||
| BC030477.1 | - | 7, 8 | 1706 | |||
| BC048831.1 | - | 6, 7 | 1388 | 4504495 | ||
| BC063070.1 | - | 7, 8 | 1569 | 5687975 | ||

Wscd1
Genomic Map