siRNA Details
| Gene Symbol: Wscd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177618.4 | NP_808286.2 | 6 | 1267 | ||
| GenBank mRNA | BC030477.1 | - | 6 | 1404 | ||
| BC048831.1 | - | 5 | 1086 | 4504495 | ||
| BC063070.1 | - | 6 | 1267 | 5687975 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wscd1| Gene Symbol: Wscd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177618.4 | NP_808286.2 | 6 | 1267 | ||
| GenBank mRNA | BC030477.1 | - | 6 | 1404 | ||
| BC048831.1 | - | 5 | 1086 | 4504495 | ||
| BC063070.1 | - | 6 | 1267 | 5687975 | ||