siRNA Details
| Gene Symbol: Spns2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276383.1 | NP_001263312.1 | 4 | 384 | ||
| NM_153060.3 | NP_694700.2 | 4 | 732 | |||
| GenBank mRNA | AB441166.1 | - | 4 | 604 | ||
| AK156007.1 | - | 5 | 378 | |||
| BC011467.1 | - | 4 | 732 | |||

Spns2
Genomic Map