siRNA Details
| Gene Symbol: Taok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144825.2 | NP_659074.2 | 2, 3 | 840 | ||
| GenBank mRNA | AK138443.1 | - | 812 | |||
| AK143530.1 | - | 2, 3 | 840 | |||
| BC047271.1 | - | 1, 2 | 187 | |||
| BC151012.1 | - | 2, 3 | 634 | 9087925 | ||
| BC151015.1 | - | 2, 3 | 634 | 9087928 | ||
| BC151016.1 | - | 2, 3 | 633 | 9087929 | ||
| CN717636.1 | - | 1, 2 | 194 | |||
| Gene Symbol: Ets1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | X53953.1 | - | 110 | |||

Taok1
Genomic Map