siRNA Details
| Gene Symbol: Utp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144826.3 | NP_659075.1 | 8 | 667 | ||
| GenBank mRNA | AF334387.1 | - | 8 | 994 | ||
| AK029151.1 | - | 8 | 668 | |||
| AK084976.1 | - | 8 | 660 | |||
| AK164010.1 | - | 8 | 672 | |||
| AK169785.1 | - | 8 | 674 | |||
| BC018250.1 | - | 8 | 631 | 4924739 | ||
| BC023920.1 | - | 8 | 661 | 5323367 | ||
| BC024850.1 | - | 8 | 662 | 5360921 | ||

Utp6
Genomic Map