siRNA Details
Gene Symbol: Utp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144826.3 | NP_659075.1 | 3, 4 | 292 | ||
GenBank mRNA | AF334387.1 | - | 3, 4 | 619 | ||
AK029151.1 | - | 3, 4 | 293 | |||
AK084976.1 | - | 3, 4 | 285 | |||
AK164010.1 | - | 3, 4 | 297 | |||
AK169785.1 | - | 3, 4 | 299 | |||
BC018250.1 | - | 3, 4 | 256 | 4924739 | ||
BC023920.1 | - | 3, 4 | 286 | 5323367 | ||
BC024850.1 | - | 3, 4 | 287 | 5360921 |
