siRNA Details
| Gene Symbol: Heatr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145432.3 | NP_663407.2 | 19 | 2973 | ||
| GenBank mRNA | AK076219.1 | - | 19 | 3102 | ||
| AK132494.1 | - | 19 | 2912 | |||
| AK136845.1 | - | 3 | 324 | |||
| AK138219.1 | - | 3 | 231 | |||
| BC003942.1 | - | 12 | 1688 | 3487155 | ||
| BC065092.1 | - | 19 | 2899 | 6855410 | ||

Heatr6
Genomic Map