siRNA Details
| Gene Symbol: Heatr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145432.3 | NP_663407.2 | 17 | 2668 | ||
| GenBank mRNA | AK076219.1 | - | 17 | 2797 | ||
| AK132494.1 | - | 17 | 2607 | |||
| AK136845.1 | - | 1 | 19 | |||
| BC003942.1 | - | 10 | 1383 | 3487155 | ||
| BC065092.1 | - | 17 | 2594 | 6855410 | ||

Heatr6
Genomic Map