siRNA Details
| Gene Symbol: Tada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172562.3 | NP_766150.1 | 14, 15 | 1198 | ||
| GenBank mRNA | AK083467.1 | - | 3, 4 | 229 | ||
| AK157938.1 | - | 14, 15 | 1191 | |||
| BC005610.1 | - | 11, 12 | 924 | |||
| BC025448.1 | - | 9, 10 | 690 | |||
| BC038821.1 | - | 14, 15 | 1201 | 3990023 | ||

Tada2a
Genomic Map