siRNA Details
| Gene Symbol: Utp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013375.1 | NP_001013393.1 | 4 | 704 | ||
| GenBank mRNA | AK032265.1 | - | 4 | 648 | ||
| AK154637.1 | - | 4 | 644 | |||
| AK167248.1 | - | 4 | 703 | |||
| BC119621.1 | - | 4 | 677 | 40062238 | ||
| BC119622.1 | - | 4 | 677 | 40062245 | ||

Utp18
Genomic Map