siRNA Details
| Gene Symbol: Kat7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195003.1 | NP_001181932.1 | 12, 13 | 1666 | ||
| NM_001195004.1 | NP_001181933.1 | 11, 12 | 1576 | |||
| NM_177619.3 | NP_808287.1 | 10, 11 | 1399 | |||
| GenBank mRNA | AK147968.1 | - | 11, 12 | 1393 | ||
| AK169458.1 | - | 8, 9 | 1583 | |||
| AK170389.1 | - | 11, 12 | 1523 | |||
| AK193799.1 | - | 2, 3 | 124 | |||
| AK197830.1 | - | 2, 3 | 124 | |||
| AK203619.1 | - | 2, 3 | 124 | |||
| AK205260.1 | - | 2, 3 | 124 | |||
| AK211586.1 | - | 2, 3 | 132 | |||
| BC024092.1 | - | 10, 11 | 1239 | |||
| BC048904.1 | - | 10, 11 | 1354 | 6492124 | ||
| BC057102.1 | - | 11, 12 | 1504 | 6401353 | ||
| DQ076247.1 | - | 12, 13 | 1594 | |||

Kat7
Genomic Map