siRNA Details
| Gene Symbol: Nr1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145434.4 | NP_663409.2 | 6 | 1942 | ||
| GenBank mRNA | AK041047.1 | - | 6 | 1798 | ||
| AK053029.1 | - | 1 | 828 | |||
| AK135640.1 | - | 6 | 1804 | |||
| AK137398.1 | - | 6 | 1829 | |||
| AK137582.1 | - | 6 | 1807 | |||
| AK154931.1 | - | 6 | 1943 | |||
| AK155597.1 | - | 6 | 1943 | |||
| BC008989.1 | - | 6 | 1387 | 3585478 | ||

Nr1d1
Genomic Map