siRNA Details
| Gene Symbol: Nr1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145434.4 | NP_663409.2 | 4 | 1166 | ||
| GenBank mRNA | AF291821.1 | - | 4 | 990 | ||
| AK041047.1 | - | 4 | 1022 | |||
| AK053029.1 | - | 1 | 2272 | |||
| AK135640.1 | - | 4 | 1028 | |||
| AK137398.1 | - | 4 | 1053 | |||
| AK137582.1 | - | 4 | 1031 | |||
| AK146430.1 | - | 4 | 1123 | |||
| AK154931.1 | - | 4 | 1167 | |||
| AK155597.1 | - | 4 | 1167 | |||
| BC008989.1 | - | 4 | 611 | 3585478 | ||

Nr1d1
Genomic Map