siRNA Details
| Gene Symbol: Atxn7l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098836.1 | NP_001092306.1 | 7, 8 | 842 | ||
| NM_001098837.1 | NP_001092307.1 | 7, 8 | 821 | |||
| GenBank mRNA | AK090368.1 | - | 7, 8 | 823 | ||
| BC059880.1 | - | 2 | 237 | |||
| BC130269.1 | - | 6, 7 | 592 | |||
| BM950535.1 | - | 5, 6 | 399 | |||

Atxn7l3
Genomic Map