siRNA Details
| Gene Symbol: March10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172568.2 | NP_766156.2 | 12 | 2693 | ||
| GenBank mRNA | AK077168.1 | - | 6 | 1313 | ||
| BC117499.1 | - | 6 | 1307 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

March10| Gene Symbol: March10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172568.2 | NP_766156.2 | 12 | 2693 | ||
| GenBank mRNA | AK077168.1 | - | 6 | 1313 | ||
| BC117499.1 | - | 6 | 1307 | |||