siRNA Details
Gene Symbol: Gpr142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181749.1 | NP_861414.1 | 3 | 563 | ||
GenBank mRNA | AY288428.1 | - | 3 | 563 | ||
AY957501.1 | - | 3 | 581 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpr142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181749.1 | NP_861414.1 | 3 | 563 | ||
GenBank mRNA | AY288428.1 | - | 3 | 563 | ||
AY957501.1 | - | 3 | 581 |