siRNA Details
| Gene Symbol: Slc16a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080934.1 | NP_001074403.1 | 3 | 518 | ||
| GenBank mRNA | AK037416.1 | - | 2 | 630 | ||
| BC172170.1 | - | 3 | 533 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc16a5| Gene Symbol: Slc16a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080934.1 | NP_001074403.1 | 3 | 518 | ||
| GenBank mRNA | AK037416.1 | - | 2 | 630 | ||
| BC172170.1 | - | 3 | 533 | |||