siRNA Details
Gene Symbol: Unk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286006.1 | NP_001272935.1 | 11 | 1450 | ||
NM_172569.4 | NP_766157.1 | 11 | 1489 | |||
GenBank mRNA | AK046395.1 | - | 11 | 1484 | ||
AK077635.1 | - | 11 | 1491 | |||
AK078095.1 | - | 11 | 1441 | |||
AK129438.1 | - | 11 | 1467 | |||
AK134361.1 | - | 11 | 1570 | |||
BC003195.1 | - | 8 | 935 | |||
BC039964.1 | - | 11 | 1451 | |||
BC054452.1 | - | 11 | 1426 | 6491704 |
