siRNA Details
| Gene Symbol: Unk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286006.1 | NP_001272935.1 | 13 | 1817 | ||
| NM_172569.4 | NP_766157.1 | 13 | 1856 | |||
| GenBank mRNA | AK046395.1 | - | 13 | 1851 | ||
| AK077635.1 | - | 13 | 1858 | |||
| AK078095.1 | - | 13 | 1808 | |||
| AK129438.1 | - | 13 | 1834 | |||
| AK134361.1 | - | 13 | 1937 | |||
| BC003195.1 | - | 10 | 1302 | |||
| BC033048.1 | - | 1 | 56 | |||
| BC039964.1 | - | 13 | 1818 | |||
| BC054452.1 | - | 13 | 1793 | 6491704 | ||

Unk
Genomic Map