siRNA Details
| Gene Symbol: Srp68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146032.3 | NP_666144.3 | 16 | 1774 | ||
| GenBank mRNA | AK033010.1 | - | 16 | 1693 | ||
| AK076003.1 | - | 16 | 1671 | |||
| AK210149.1 | - | 2 | 79 | |||
| AK211431.1 | - | 2 | 80 | |||
| BC033573.1 | - | 12 | 1074 | 5323201 | ||
| BC070422.1 | - | 16 | 1774 | 6403747 | ||

Srp68
Genomic Map