siRNA Details
| Gene Symbol: Ube2o | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173755.3 | NP_776116.2 | 18 | 3251 | ||
| GenBank mRNA | AK035970.1 | - | 1 | 470 | ||
| AK045398.1 | - | 5 | 621 | |||
| AK129431.1 | - | 18 | 3251 | |||
| BC023329.1 | - | 5 | 981 | |||
| BC039233.1 | - | 10 | 1717 | |||
| BC051108.1 | - | 9 | 1596 | |||
| BC055067.1 | - | 4 | 343 | |||
| BC059193.1 | - | 18 | 2876 | 4160667 | ||

Ube2o
Genomic Map